accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH2N

CAS RN 15760-35-7

IUPAC Name: 3-methylidenecyclobutane-1-carbonitrile
Synonyms: 3-methylidenecyclobutane-1-carbonitrile
Molecular Weight (g/mol): 93.13
Molecular Formula: C6H7N
InChi Key: ZRWMAMOBIQQJSA-UHFFFAOYSA-N
SMILES: C=C1CC(C1)C#N
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)