accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 157610-82-7

IUPAC Name: [(isoquinolin-4-yl)methyl](methyl)amine
Synonyms: (isoquinolin-4-ylmethyl)(methyl)amine
Molecular Weight (g/mol): 172.23
Molecular Formula: C11H12N2
InChi Key: PISGLKXJHQKIFN-UHFFFAOYSA-N
SMILES: CNCC1=C2C=CC=CC2=CN=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results