accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 15764-52-0

IUPAC Name: 2-(2-hydroxyphenoxy)phenol
Synonyms: 2-(2-hydroxyphenoxy)phenol
Molecular Weight (g/mol): 202.21
Molecular Formula: C12H10O3
InChi Key: VXHYVVAUHMGCEX-UHFFFAOYSA-N
SMILES: OC1=CC=CC=C1OC1=CC=CC=C1O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results