accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 157688-46-5

IUPAC Name: 2-{1-[(tert-butoxy)carbonyl]piperidin-4-yl}acetate
Synonyms: 2-[1-(tert-butoxycarbonyl)piperidin-4-yl]acetate
Molecular Weight (g/mol): 242.3
Molecular Formula: C12H20NO4
InChi Key: ZXFLMSIMHISJFV-UHFFFAOYSA-M
SMILES: CC(C)(C)OC(=O)N1CCC(CC([O-])=O)CC1
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)