accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CBrBr

CAS RN 157761-91-6

IUPAC Name: 1,3-dibromo-5-tetradecylbenzene
Synonyms: 1,3-dibromo-5-tetradecylbenzene
Molecular Weight (g/mol): 432.28
Molecular Formula: C20H32Br2
InChi Key: DEEYQDRVVZEMDY-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCC1=CC(Br)=CC(Br)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results