accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHONHN

CAS RN 15788-16-6

IUPAC Name: 1H-1,3-benzodiazole-6-carboxylic acid
Synonyms: 3H-1,3-benzodiazole-5-carboxylic acid
Molecular Weight (g/mol): 162.15
Molecular Formula: C8H6N2O2
InChi Key: COYPLDIXZODDDL-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=C2N=CNC2=C1
Percent Purity 
  • (2)
  • (1)
  • (6)
Quantity 
  • (1)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (2)
  • (6)
  • (2)
  • (1)
  • (6)
  • (1)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)