accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOHNOH

CAS RN 157887-82-6

IUPAC Name: (9H-fluoren-9-yl)methyl N-(3-hydroxypropyl)carbamate
Synonyms: 9H-fluoren-9-ylmethyl N-(3-hydroxypropyl)carbamate
Molecular Weight (g/mol): 297.35
Molecular Formula: C18H19NO3
InChi Key: GNXZNUJDAMSJFZ-UHFFFAOYSA-N
SMILES: OCCCNC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results