accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 157911-55-2

IUPAC Name: 1-(bromomethyl)-2,3,4-trifluorobenzene
Synonyms: 1-(bromomethyl)-2,3,4-trifluorobenzene
Molecular Weight (g/mol): 225.01
Molecular Formula: C7H4BrF3
InChi Key: DGSXDQVPGXFOAN-UHFFFAOYSA-N
SMILES: FC1=CC=C(CBr)C(F)=C1F
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results