accessibility menu, dialog, popup

UserName

Viewing profile as user:
NNNOOH

CAS RN 158014-74-5

IUPAC Name: 4-[6-(pyridin-2-yl)-[2,2'-bipyridin]-4-yl]benzoic acid
Synonyms: 4-[6-(pyridin-2-yl)-[2,2'-bipyridin]-4-yl]benzoic acid
Molecular Weight (g/mol): 353.38
Molecular Formula: C22H15N3O2
InChi Key: RPRRPKJKACRJDD-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=C(C=C1)C1=CC(=NC(=C1)C1=CC=CC=N1)C1=CC=CC=N1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results