accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 15802-00-3

H3CNCH3H3CH3CBrIBr

CAS RN 15802-00-3

IUPAC Name: dibromoiodanuide; tetrabutylazanium
Synonyms: dibromoiodanuide; tetrabutylammonium ion
Molecular Weight (g/mol): 529.18
Molecular Formula: C16H36Br2IN
InChi Key: LFZGDXLUTJIZAG-UHFFFAOYSA-N
SMILES: Br[I-]Br.CCCC[N+](CCCC)(CCCC)CCCC
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results