accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3ONHNFFFFF

CAS RN 1580464-60-3

IUPAC Name: N-[6-(1,1,2,2,2-pentafluoroethyl)pyridin-3-yl]acetamide
Synonyms: N-[6-(1,1,2,2,2-pentafluoroethyl)pyridin-3-yl]acetamide
Molecular Weight (g/mol): 254.16
Molecular Formula: C9H7F5N2O
InChi Key: FTIYUAIKVLBBTH-UHFFFAOYSA-N
SMILES: CC(=O)NC1=CN=C(C=C1)C(F)(F)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results