accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CH3CCH3OONHNFFFFF

CAS RN 1580464-68-1

IUPAC Name: tert-butyl N-[5-(1,1,2,2,2-pentafluoroethyl)pyridin-2-yl]carbamate
Synonyms: tert-butyl N-[5-(1,1,2,2,2-pentafluoroethyl)pyridin-2-yl]carbamate
Molecular Weight (g/mol): 312.24
Molecular Formula: C12H13F5N2O2
InChi Key: ZASNKMWFNLRBGM-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)NC1=NC=C(C=C1)C(F)(F)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results