accessibility menu, dialog, popup

UserName

Viewing profile as user:
NFFFOOH

CAS RN 158063-66-2

IUPAC Name: 4-(trifluoromethyl)pyridine-3-carboxylic acid
Synonyms: 4-(trifluoromethyl)pyridine-3-carboxylic acid
Molecular Weight (g/mol): 191.11
Molecular Formula: C7H4F3NO2
InChi Key: LMRJHNFECNKDKH-UHFFFAOYSA-N
SMILES: OC(=O)C1=C(C=CN=C1)C(F)(F)F
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (3)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (5)
  • (3)
  • (3)
  • (3)
  • (2)
  • (1)