accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOO(S)OPOHOONHOO

CAS RN 158171-14-3

IUPAC Name: (2S)-3-{[(benzyloxy)(hydroxy)phosphoryl]oxy}-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)propanoic acid
Synonyms: (2S)-3-{[benzyloxy(hydroxy)phosphoryl]oxy}-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}propanoic acid
Molecular Weight (g/mol): 497.44
Molecular Formula: C25H24NO8P
InChi Key: ZBPUWGDUVAAWJY-QHCPKHFHSA-N
SMILES: OC(=O)[C@H](COP(O)(=O)OCC1=CC=CC=C1)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (2)
  • (3)