accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOHOOHONH(S)(S)H3CCH3

CAS RN 158257-40-0

IUPAC Name: (3S,4S)-4-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3-hydroxy-6-methylheptanoic acid
Synonyms: (3S,4S)-4-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}-3-hydroxy-6-methylheptanoic acid
Molecular Weight (g/mol): 397.47
Molecular Formula: C23H27NO5
InChi Key: JGUYYODGVQXZAY-SFTDATJTSA-N
SMILES: CC(C)C[C@H](NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2)[C@@H](O)CC(O)=O
Grade 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)