accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 158271-95-5

IUPAC Name: 1-[(1R)-2-hydroxy-1-phenylethyl]-2,5-dihydro-1H-pyrrol-2-one
Synonyms: 1-[(1R)-2-hydroxy-1-phenylethyl]-5H-pyrrol-2-one
Molecular Weight (g/mol): 203.24
Molecular Formula: C12H13NO2
InChi Key: IPPHIZDKNVGDJX-NSHDSACASA-N
SMILES: OC[C@H](N1CC=CC1=O)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results