accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOFOO

CAS RN 1583-67-1

IUPAC Name: 3-fluorobenzene-1,2-dicarboxylate
Synonyms: 3-fluorophthalate
Molecular Weight (g/mol): 182.11
Molecular Formula: C8H3FO4
InChi Key: BBCQSMSCEJBIRD-UHFFFAOYSA-L
SMILES: [O-]C(=O)C1=CC=CC(F)=C1C([O-])=O
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)