accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOBr

CAS RN 15852-73-0

IUPAC Name: (3-bromophenyl)methanol
Synonyms: (3-bromophenyl)methanol
Molecular Weight (g/mol): 187.04
Molecular Formula: C7H7BrO
InChi Key: FSWNRRSWFBXQCL-UHFFFAOYSA-N
SMILES: OCC1=CC=CC(Br)=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results