accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COONNNNNNCH3OCH3OH3C

CAS RN 1585236-34-5

IUPAC Name: 5,6-bis(azidomethyl)-2,3-dimethoxy-2,3-dimethyl-1,4-dioxane
Synonyms: 5,6-bis(azidomethyl)-2,3-dimethoxy-2,3-dimethyl-1,4-dioxane
Molecular Weight (g/mol): 286.29
Molecular Formula: C10H18N6O4
InChi Key: VKZQUTPIKFBBMZ-UHFFFAOYNA-N
SMILES: COC1(C)OC(CN=[N+]=[N-])C(CN=[N+]=[N-])OC1(C)OC
Percent Purity 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results