CAS RN 15854-08-7
CAS RN 15854-08-7
IUPAC Name:
ethyl 2-amino-4-(4-methylphenyl)thiophene-3-carboxylate
Synonyms:
ethyl 2-amino-4-(4-methylphenyl)thiophene-3-carboxylate
Molecular Weight (g/mol):
261.34
Molecular Formula:
C14H15NO2S
InChi Key:
GOQSMGNEJVWZIG-UHFFFAOYSA-N
SMILES:
CCOC(=O)C1=C(N)SC=C1C1=CC=C(C)C=C1
1
–
1
of
1
results
1
Sigma Aldrich Ethyl 2-amino-4-p-tolylthiophene-3-carboxylate
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.