accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1586-92-1

H3COAlCH3CH3

CAS RN 1586-92-1

IUPAC Name: ethoxydiethylalumane
Synonyms: diethylaluminium ethoxide
Molecular Weight (g/mol): 130.17
Molecular Formula: C6H15AlO
InChi Key: GCPCLEKQVMKXJM-UHFFFAOYSA-N
SMILES: CCO[Al](CC)CC
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Boiling Point 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)