CAS RN 15862-18-7
IUPAC Name:
5,5'-dibromo-2,2'-bipyridine
Synonyms:
5,5'-dibromo-2,2'-bipyridine
Molecular Weight (g/mol):
313.98
Molecular Formula:
C10H6Br2N2
InChi Key:
JNWPRPLNUUMYCM-UHFFFAOYSA-N
SMILES:
BrC1=CN=C(C=C1)C1=NC=C(Br)C=C1
1
–
1
of
1
results
1
5,5'-Dibromo-2,2'-bipyridyl 98.0+%, TCI America™
CAS: 15862-18-7 Molecular Formula: C10H6Br2N2 Molecular Weight (g/mol): 313.98 MDL Number: MFCD06657883 InChI Key: JNWPRPLNUUMYCM-UHFFFAOYSA-N Synonym: 5,5′C-Dibromo-2,2′C-bipyridine PubChem CID: 10903103 IUPAC Name: 5-bromo-2-(5-bromopyridin-2-yl)pyridine SMILES: C1=CC(=NC=C1Br)C2=NC=C(C=C2)Br