accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONOHNOBr

CAS RN 15862-34-7

IUPAC Name: 5-bromo-3-nitro-1,2-dihydropyridin-2-one
Synonyms: 5-bromo-3-nitro-1H-pyridin-2-one
Molecular Weight (g/mol): 218.99
Molecular Formula: C5H3BrN2O3
InChi Key: WXRLCVUDLFFTFF-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC(Br)=CNC1=O
Grade 
  • (2)
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results