accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 15864-32-1

IUPAC Name: 6-bromo-1,3-benzothiazol-2-amine
Synonyms: 6-bromo-1,3-benzothiazol-2-amine
Molecular Weight (g/mol): 229.1
Molecular Formula: C7H5BrN2S
InChi Key: VZEBSJIOUMDNLY-UHFFFAOYSA-N
SMILES: NC1=NC2=CC=C(Br)C=C2S1
Molecular Weight (g/mol) 
  • (1)
  • (6)
Percent Purity 
  • (1)
  • (2)
  • (1)
  • (3)
Quantity 
  • (2)
  • (2)
  • (3)
Physical Form 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (1)
  • (5)
  • (1)
  • (6)
  • (1)
  • (2)
  • (1)
  • (3)
  • (2)
  • (2)
  • (3)
  • (1)
  • (2)