accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 158663-69-5

IUPAC Name: piperazine-2-carboxylic acid dihydrochloride
Synonyms: piperazine-2-carboxylic acid dihydrochloride
Molecular Weight (g/mol): 203.06
Molecular Formula: C5H12Cl2N2O2
InChi Key: WNSDZBQLMGKPQS-UHFFFAOYNA-N
SMILES: Cl.Cl.OC(=O)C1CNCCN1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results