accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOOHOOH3COOH3C

CAS RN 1587-20-8

IUPAC Name: 1,2,3-trimethyl 2-hydroxypropane-1,2,3-tricarboxylate
Synonyms: 1,2,3-trimethyl 2-hydroxypropane-1,2,3-tricarboxylate
Molecular Weight (g/mol): 234.2
Molecular Formula: C9H14O7
InChi Key: HDDLVZWGOPWKFW-UHFFFAOYSA-N
SMILES: COC(=O)CC(O)(CC(=O)OC)C(=O)OC
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)