accessibility menu, dialog, popup

UserName

Viewing profile as user:
HClNH2NOO

CAS RN 15873-51-5

IUPAC Name: hydrogen 4-nitroaniline chloride
Synonyms: hydrogen p-nitroaniline chloride
Molecular Weight (g/mol): 174.58
Molecular Formula: C6H7ClN2O2
InChi Key: LNJUVOPKIUQOQK-UHFFFAOYSA-N
SMILES: [H+].[Cl-].NC1=CC=C(C=C1)[N+]([O-])=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results