accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 15894-04-9

IUPAC Name: (4-fluorophenyl)methanethiol
Synonyms: (4-fluorophenyl)methanethiol
Molecular Weight (g/mol): 142.19
Molecular Formula: C7H7FS
InChi Key: RKTRHMNWVZRZJQ-UHFFFAOYSA-N
SMILES: FC1=CC=C(CS)C=C1
Boiling Point 
  • (3)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (3)
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (3)
  • (1)
  • (3)
  • (2)
  • (2)
  • (1)