accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CCH3OONNOH

CAS RN 158985-37-6

IUPAC Name: tert-butyl 4-[4-(hydroxymethyl)phenyl]piperazine-1-carboxylate
Synonyms: tert-butyl 4-[4-(hydroxymethyl)phenyl]piperazine-1-carboxylate
Molecular Weight (g/mol): 292.38
Molecular Formula: C16H24N2O3
InChi Key: PXKYXTXYWJKTSQ-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)N1CCN(CC1)C1=CC=C(CO)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results