accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CO

CAS RN 1590-08-5

IUPAC Name: 2-methyl-1,2,3,4-tetrahydronaphthalen-1-one
Synonyms: 2-methyl-3,4-dihydro-2H-naphthalen-1-one
Molecular Weight (g/mol): 160.22
Molecular Formula: C11H12O
InChi Key: GANIBVZSZGNMNB-UHFFFAOYNA-N
SMILES: CC1CCC2=CC=CC=C2C1=O
Boiling Point 
  • (1)
  • (1)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (1)
  • (1)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)