accessibility menu, dialog, popup

UserName

Viewing profile as user:
I

CAS RN 1591-31-7

IUPAC Name: 4-iodo-1,1'-biphenyl
Synonyms: 1,1'-biphenyl, 4-iodo-
Molecular Weight (g/mol): 280.11
Molecular Formula: C12H9I
InChi Key: NXYICUMSYKIABQ-UHFFFAOYSA-N
SMILES: IC1=CC=C(C=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (1)
  • (7)
Percent Purity 
  • (2)
  • (4)
  • (2)
Quantity 
  • (2)
  • (1)
  • (2)
  • (3)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (2)
  • (5)
  • (1)
  • (7)
  • (2)
  • (4)
  • (2)
  • (2)
  • (1)
  • (2)
  • (3)
  • (4)
  • (2)