accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNOH3COH3CONHO

CAS RN 159109-11-2

IUPAC Name: 5-ethyl-7,8-dimethoxy-1H,2H,3H-pyrrolo[3,4-c]isoquinoline-1,3-dione
Synonyms: 5-ethyl-7,8-dimethoxy-2H-pyrrolo[3,4-c]isoquinoline-1,3-dione
Molecular Weight (g/mol): 286.29
Molecular Formula: C15H14N2O4
InChi Key: ULVWJFBHQIXEPE-UHFFFAOYSA-N
SMILES: CCC1=C2C=C(OC)C(OC)=CC2=C2C(=O)NC(=O)C2=N1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results