accessibility menu, dialog, popup

UserName

Viewing profile as user:
BOSiCH3H3CH3CCH3H3CHOOH

CAS RN 159191-56-7

IUPAC Name: {4-[(tert-butyldimethylsilyl)oxy]phenyl}boronic acid
Synonyms: 4-[(tert-butyldimethylsilyl)oxy]phenylboronic acid
Molecular Weight (g/mol): 252.19
Molecular Formula: C12H21BO3Si
InChi Key: NVHHEADQQACSCJ-UHFFFAOYSA-N
SMILES: CC(C)(C)[Si](C)(C)OC1=CC=C(C=C1)B(O)O
Molecular Weight (g/mol) 
  • (1)
  • (1)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results