accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1592-12-7

IUPAC Name: (4-ethenylphenyl)methyl acetate
Synonyms: (4-ethenylphenyl)methyl acetate
Molecular Weight (g/mol): 176.22
Molecular Formula: C11H12O2
InChi Key: LEIKPUSDAWATBV-UHFFFAOYSA-N
SMILES: CC(=O)OCC1=CC=C(C=C)C=C1
Boiling Point 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)