CAS RN 1592-43-4
IUPAC Name:
1',3',3'-trimethyl-1',3'-dihydrospiro[benzo[f]chromene-3,2'-indole]
Synonyms:
1',3',3'-trimethylspiro[benzo[f]chromene-3,2'-indole]
Molecular Weight (g/mol):
327.43
Molecular Formula:
C23H21NO
InChi Key:
DTQKEQFXLWFVCS-UHFFFAOYNA-N
SMILES:
CN1C2=CC=CC=C2C(C)(C)C11OC2=CC=C3C=CC=CC3=C2C=C1
1
–
1
of
1
results
1
1,3,3-Trimethylindolino-beta-naphthopyrylospiran 98.0+%, TCI America™
CAS: 1592-43-4 Molecular Formula: C23H21NO Molecular Weight (g/mol): 327.427 MDL Number: MFCD00059909 InChI Key: DTQKEQFXLWFVCS-UHFFFAOYSA-N PubChem CID: 2728827 IUPAC Name: 1',3',3'-trimethylspiro[benzo[f]chromene-3,2'-indole] SMILES: CC1(C2=CC=CC=C2N(C13C=CC4=C(O3)C=CC5=CC=CC=C54)C)C