accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 15922-01-7

IUPAC Name: potassium 4-nitrobenzoate
Synonyms: potassium 4-nitrobenzoate
Molecular Weight (g/mol): 205.21
Molecular Formula: C7H4KNO4
InChi Key: MKBKSBKMELRIKB-UHFFFAOYSA-M
SMILES: [K+].[O-]C(=O)C1=CC=C(C=C1)[N+]([O-])=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results