accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 159275-17-9

IUPAC Name: benzyl 4-(bromomethyl)piperidine-1-carboxylate
Synonyms: benzyl 4-(bromomethyl)piperidine-1-carboxylate
Molecular Weight (g/mol): 312.21
Molecular Formula: C14H18BrNO2
InChi Key: XJHKDSZGAWXUTB-UHFFFAOYSA-N
SMILES: BrCC1CCN(CC1)C(=O)OCC1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results