accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SiH3CNHSiCH3H3C

CAS RN 15933-59-2

IUPAC Name: —
Synonyms: —
Molecular Weight (g/mol): 131.33
Molecular Formula: C4H13NSi2
InChi Key: GJWAPAVRQYYSTK-UHFFFAOYSA-N
SMILES: C[Si](C)N[Si](C)C
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)