accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NNOHO

CAS RN 159589-70-5

IUPAC Name: 2-(4-methylpiperazin-1-yl)benzoic acid
Synonyms: 2-(4-methylpiperazin-1-yl)benzoic acid
Molecular Weight (g/mol): 220.27
Molecular Formula: C12H16N2O2
InChi Key: WKGFDTBUUBBWJZ-UHFFFAOYSA-N
SMILES: CN1CCN(CC1)C1=CC=CC=C1C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results