accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 159589-71-6

CH3NNOOH

CAS RN 159589-71-6

IUPAC Name: 2-(2-methyl-1H-imidazol-1-yl)benzoic acid
Synonyms: 2-(2-methylimidazol-1-yl)benzoic acid
Molecular Weight (g/mol): 202.21
Molecular Formula: C11H10N2O2
InChi Key: PDOVVVXGESUKQF-UHFFFAOYSA-N
SMILES: CC1=NC=CN1C1=CC=CC=C1C(O)=O
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results