accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOHH2NFFF

CAS RN 15959-93-0

IUPAC Name: 2-amino-4,4,4-trifluorobutanoic acid
Synonyms: 2-amino-4,4,4-trifluorobutanoic acid
Molecular Weight (g/mol): 157.09
Molecular Formula: C4H6F3NO2
InChi Key: AQPCXCOPDSEKQT-UHFFFAOYNA-N
SMILES: NC(CC(F)(F)F)C(O)=O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)