accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClClH2N(S)HONHNHH

CAS RN 1596-64-1

IUPAC Name: dihydrogen (2S)-2-amino-3-(1H-imidazol-5-yl)propan-1-ol dichloride
Synonyms: dihydrogen HSO dichloride
Molecular Weight (g/mol): 214.09
Molecular Formula: C6H13Cl2N3O
InChi Key: FRCAFNBBXRWXQA-XRIGFGBMSA-N
SMILES: [H+].[H+].[Cl-].[Cl-].N[C@H](CO)CC1=CN=CN1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)