accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 159622-10-3

IUPAC Name: 1-{[(tert-butoxy)carbonyl]amino}-2-ethenylcyclopropane-1-carboxylic acid
Synonyms: 1-[(tert-butoxycarbonyl)amino]-2-ethenylcyclopropane-1-carboxylic acid
Molecular Weight (g/mol): 227.26
Molecular Formula: C11H17NO4
InChi Key: RFAQWADNTLIWMG-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)NC1(CC1C=C)C(O)=O
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)