accessibility menu, dialog, popup

UserName

Viewing profile as user:
NaNaOOOO

CAS RN 15968-01-1

IUPAC Name: disodium benzene-1,2-dicarboxylate
Synonyms: disodium phthalate
Molecular Weight (g/mol): 210.1
Molecular Formula: C8H4Na2O4
InChi Key: HQWKKEIVHQXCPI-UHFFFAOYSA-L
SMILES: [Na+].[Na+].[O-]C(=O)C1=CC=CC=C1C([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results