accessibility menu, dialog, popup

UserName

Viewing profile as user:
SFFFFFClN

CAS RN 159727-28-3

IUPAC Name: 3-chloro-5-(pentafluoro-λ⁶-sulfanyl)benzonitrile
Synonyms: 3-chloro-5-(pentafluoro-λ⁶-sulfanyl)benzonitrile
Molecular Weight (g/mol): 263.61
Molecular Formula: C7H3ClF5NS
InChi Key: HASUULAKVNPSHK-UHFFFAOYSA-N
SMILES: FS(F)(F)(F)(F)C1=CC(Cl)=CC(=C1)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results