accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 15980-15-1

IUPAC Name: 1,4-oxathiane
Synonyms: 1,4-oxathiane
Molecular Weight (g/mol): 104.17
Molecular Formula: C4H8OS
InChi Key: JBYHSSAVUBIJMK-UHFFFAOYSA-N
SMILES: C1CSCCO1
Boiling Point 
  • (1)
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (3)
  • (1)
Percent Purity 
  • (2)
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)