accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 15989-47-6

IUPAC Name: 3-(diaminomethylidene)-1-phenylthiourea
Synonyms: 3-(diaminomethylidene)-1-phenylthiourea
Molecular Weight (g/mol): 194.26
Molecular Formula: C8H10N4S
InChi Key: IKGVILAOZDBXSV-UHFFFAOYSA-N
SMILES: NC(N)=NC(=S)NC1=CC=CC=C1
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results