accessibility menu, dialog, popup

UserName

Viewing profile as user:
BOOHHOOH(E/Z)

CAS RN 159896-15-8

IUPAC Name: 3-[4-(dihydroxyboranyl)phenyl]prop-2-enoic acid
Synonyms: 3-[4-(dihydroxyboranyl)phenyl]prop-2-enoic acid
Molecular Weight (g/mol): 191.98
Molecular Formula: C9H9BO4
InChi Key: IEMLKNHGGSYOMP-UHFFFAOYSA-N
SMILES: OB(O)C1=CC=C(C=CC(O)=O)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results