accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1599-67-3

H3CCH2

CAS RN 1599-67-3

IUPAC Name: docos-1-ene
Synonyms: 1-docosene C20-24 olefin
Molecular Weight (g/mol): 308.59
Molecular Formula: C22H44
InChi Key: SPURMHFLEKVAAS-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCCCCCCC=C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results