accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 160017-09-4

IUPAC Name: 5-(pyrrolidin-1-yl)pyridine-2-carbonitrile
Synonyms: 5-(pyrrolidin-1-yl)pyridine-2-carbonitrile
Molecular Weight (g/mol): 173.22
Molecular Formula: C10H11N3
InChi Key: WBEGYGZFOINKIL-UHFFFAOYSA-N
SMILES: N#CC1=NC=C(C=C1)N1CCCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results